Assay Details
| Gene Symbol: fam228a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045186.2 | NP_001038651.2 | 8 - 9 | 949 | 110 | |
| XM_017352515.2 | XP_017208004.1 | 7 - 8 | 1082 | 110 | ||
| XM_021468502.1 | XP_021324177.1 | 7 - 8 | 2072 | 110 | ||
| GenBank mRNA | BC135057.1 | - | 8 - 9 | 949 | 110 | |
More Information
Set Membership: |
|

fam228a
Genomic Map