Assay Details
| Gene Symbol: oxr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045203.1 | NP_001038668.1 | 13 - 14 | 2186 | 97 | |
| XM_005159458.3 | XP_005159515.1 | 16 - 17 | 2580 | 97 | ||
| XM_005159459.3 | XP_005159516.1 | 15 - 16 | 2499 | 97 | ||
| XM_009294057.3 | XP_009292332.1 | 6 - 7 | 991 | 97 | ||
More Information
Set Membership: |
|

oxr1b
Genomic Map