Assay Details
| Gene Symbol: ccdc85ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045384.1 | NP_001038849.1 | 1 - 2 | 968 | 120 | |
| XM_005158676.4 | XP_005158733.1 | 1 - 2 | 992 | 120 | ||
| GenBank mRNA | BC122223.1 | - | 1 - 2 | 968 | 120 | |
More Information
Set Membership: |
|

ccdc85ca
Genomic Map