Assay Details
| Gene Symbol: srrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045427.1 | NP_001038892.1 | 5 - 6 | 775 | 102 | |
| XM_005172446.4 | XP_005172503.1 | 5 - 6 | 795 | 102 | ||
| XM_009303729.3 | XP_009302004.1 | 5 - 6 | 733 | 102 | ||
| XM_017357373.2 | XP_017212862.1 | 4 - 5 | 803 | 102 | ||
| XM_017357374.2 | XP_017212863.1 | 4 - 5 | 697 | 102 | ||
| GenBank mRNA | BC122414.1 | - | 5 - 6 | 775 | 102 | |
More Information
Set Membership: |
|

srrd
Genomic Map