Assay Details
| Gene Symbol: mrps5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076596.1 | NP_001070064.1 | 3 - 4 | 327 | 58 | |
| GenBank mRNA | BC124422.1 | - | 3 - 4 | 327 | 58 | |
More Information
Set Membership: |
|

mrps5
Genomic Map