Assay Details
| Gene Symbol: rgs7bpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076608.1 | NP_001070076.1 | 1 - 2 | 537 | 76 | |
| GenBank mRNA | BC124550.1 | - | 1 - 2 | 537 | 76 | |
| BC165108.1 | - | 1 - 2 | 176 | 76 | ||
More Information
Set Membership: |
|

rgs7bpa
Genomic Map