Assay Details
| Gene Symbol: cdin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076722.1 | NP_001070190.1 | 10 - 11 | 735 | 68 | |
| GenBank mRNA | BC124527.1 | - | 10 - 11 | 735 | 68 | |
More Information
Set Membership: |
|

cdin1
Genomic Map