Assay Details
| Gene Symbol: ipcef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076758.1 | NP_001070226.1 | 3 - 4 | 362 | 108 | |
| XM_021468524.1 | XP_021324199.1 | 3 - 4 | 698 | 108 | ||
| GenBank mRNA | BC124514.1 | - | 3 - 4 | 362 | 108 | |
| BC164852.1 | - | 2 - 3 | 270 | 108 | ||
More Information
Set Membership: |
|

ipcef1
Genomic Map