Assay Details
| Gene Symbol: ipcef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076758.1 | NP_001070226.1 | 4 - 5 | 433 | 82 | |
| XM_021468524.1 | XP_021324199.1 | 4 - 5 | 769 | 82 | ||
| GenBank mRNA | BC124514.1 | - | 4 - 5 | 433 | 82 | |
| BC164852.1 | - | 3 - 4 | 341 | 82 | ||
More Information
Set Membership: |
|

ipcef1
Genomic Map