Assay Details
| Gene Symbol: tfb1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076769.1 | NP_001070237.1 | 6 - 7 | 724 | 72 | |
| XM_005169880.4 | XP_005169937.1 | 6 - 7 | 738 | 72 | ||
| XM_021467233.1 | XP_021322908.1 | 6 - 7 | 796 | 72 | ||
| XM_021467235.1 | XP_021322910.1 | 5 - 6 | 647 | 72 | ||
| GenBank mRNA | BC124402.1 | - | 6 - 7 | 724 | 72 | |
| BC165534.1 | - | 5 - 6 | 701 | 72 | ||
More Information
Set Membership: |
|

tfb1m
Genomic Map