Assay Details
| Gene Symbol: tusc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077299.1 | NP_001070767.1 | 2 - 3 | 292 | 105 | |
| GenBank mRNA | BC124726.1 | - | 2 - 3 | 292 | 105 | |
| BC165126.1 | - | 1 - 2 | 183 | 105 | ||
More Information
Set Membership: |
|

tusc2a
Genomic Map