Assay Details
| Gene Symbol: tusc2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077299.1 | NP_001070767.1 | 3 - 4 | 414 | 129 | |
| GenBank mRNA | BC124726.1 | - | 3 - 4 | 414 | 129 | |
More Information
Set Membership: |
|

tusc2a
Genomic Map