Assay Details
| Gene Symbol: pdpk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077344.1 | NP_001070812.1 | 12 - 13 | 1587 | 107 | |
| GenBank mRNA | BC124736.1 | - | 12 - 13 | 1587 | 107 | |
More Information
Set Membership: |
|

pdpk1a
Genomic Map