Assay Details
| Gene Symbol: pdpk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077344.1 | NP_001070812.1 | 4 - 5 | 663 | 93 | |
| GenBank mRNA | BC091466.1 | - | 5 - 6 | 605 | 93 | |
| BC124736.1 | - | 4 - 5 | 663 | 93 | ||
More Information
Set Membership: |
|

pdpk1a
Genomic Map