Assay Details
| Gene Symbol: bicdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193591.1 | NP_001180520.1 | 5 - 6 | 996 | 63 | |
| XM_005165558.4 | XP_005165615.1 | 5 - 6 | 987 | 63 | ||
| XM_009301866.3 | XP_009300141.1 | 6 - 7 | 912 | 63 | ||
| GenBank mRNA | BC125915.1 | - | 4 - 5 | 925 | 63 | |
More Information
Set Membership: |
|

bicdl2
Genomic Map