Assay Details
| Gene Symbol: mindy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077756.1 | NP_001071224.1 | 2 - 3 | 731 | 102 | |
| GenBank mRNA | BC124766.1 | - | 2 - 3 | 731 | 102 | |
More Information
Set Membership: |
|

mindy3
Genomic Map