Assay Details
| Gene Symbol: prickle2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077783.2 | NP_001071251.2 | 3 - 4 | 604 | 128 | |
| GenBank mRNA | BC139696.1 | - | 3 - 4 | 604 | 128 | |
More Information
Set Membership: |
|

prickle2a
Genomic Map