Assay Details
| Gene Symbol: dclk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135121.1 | NP_001128593.1 | 2 - 3 | 884 | 90 | |
| NM_001145787.1 | NP_001139259.1 | 2 - 3 | 884 | 90 | ||
| NM_001145788.1 | NP_001139260.1 | 2 - 3 | 884 | 90 | ||
| NM_001145789.1 | NP_001139261.1 | 2 - 3 | 884 | 90 | ||
| XM_009292593.3 | XP_009290868.1 | 2 - 3 | 1138 | 90 | ||
| GenBank mRNA | AB266175.1 | - | 2 - 3 | 884 | 90 | |
| AB266176.1 | - | 2 - 3 | 884 | 90 | ||
| AB266177.1 | - | 2 - 3 | 884 | 90 | ||
| AB266178.1 | - | 2 - 3 | 884 | 90 | ||
| AB266179.1 | - | 2 - 3 | 884 | 90 | ||
More Information
Set Membership: |
|

dclk2a
Genomic Map