Assay Details
| Gene Symbol: dclk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135121.1 | NP_001128593.1 | 7 - 8 | 1366 | 62 | |
| NM_001145787.1 | NP_001139259.1 | 6 - 7 | 1309 | 62 | ||
| NM_001145788.1 | NP_001139260.1 | 7 - 8 | 1366 | 62 | ||
| NM_001145789.1 | NP_001139261.1 | 7 - 8 | 1357 | 62 | ||
| XM_009292593.3 | XP_009290868.1 | 7 - 8 | 1593 | 62 | ||
| GenBank mRNA | AB266175.1 | - | 7 - 8 | 1366 | 62 | |
| AB266176.1 | - | 6 - 7 | 1309 | 62 | ||
| AB266177.1 | - | 7 - 8 | 1366 | 62 | ||
| AB266178.1 | - | 7 - 8 | 1357 | 62 | ||
| AB266179.1 | - | 6 - 7 | 1309 | 62 | ||
| BC163926.1 | - | 7 - 8 | 1367 | 62 | ||
| BC168500.1 | - | 6 - 7 | 1476 | 62 | ||
More Information
Set Membership: |
|

dclk2a
Genomic Map