Assay Details
| Gene Symbol: atp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083580.1 | NP_001077049.1 | 1 - 2 | 399 | 117 | |
| GenBank mRNA | BC135073.1 | - | 1 - 2 | 399 | 117 | |
More Information
Set Membership: |
|

atp23
Genomic Map