Assay Details
| Gene Symbol: smarcc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089367.1 | NP_001082836.1 | 9 - 10 | 916 | 57 | |
| GenBank mRNA | BC095743.1 | - | 9 - 10 | 902 | 57 | |
| BC134882.1 | - | 9 - 10 | 916 | 57 | ||
More Information
Set Membership: |
|

smarcc1b
Genomic Map