Assay Details
| Gene Symbol: abhd6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328359.1 | NP_001315288.1 | 2 - 3 | 339 | 61 | |
| XM_009296744.3 | XP_009295019.1 | 2 - 3 | 576 | 61 | ||
| GenBank mRNA | BC134130.1 | - | 2 - 3 | 339 | 61 | |
| EH542669.1 | - | 2 - 3 | 324 | 61 | ||
More Information
Set Membership: |
|

abhd6b
Genomic Map