Assay Details
| Gene Symbol: abhd6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328359.1 | NP_001315288.1 | 5 - 6 | 735 | 79 | |
| XM_009296744.3 | XP_009295019.1 | 5 - 6 | 972 | 79 | ||
| GenBank mRNA | BC134130.1 | - | 5 - 6 | 735 | 79 | |
| EH542669.1 | - | 5 - 6 | 720 | 79 | ||
| EH563805.1 | - | 4 - 5 | 579 | 79 | ||
More Information
Set Membership: |
|

abhd6b
Genomic Map