Assay Details
| Gene Symbol: sp3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089498.1 | NP_001082967.1 | 3 - 4 | 1415 | 70 | |
| XM_005167417.4 | XP_005167474.1 | 4 - 5 | 1801 | 70 | ||
| XM_005167418.4 | XP_005167475.1 | 4 - 5 | 1802 | 70 | ||
| GenBank mRNA | BC139667.1 | - | 3 - 4 | 1415 | 70 | |
More Information
Set Membership: |
|

sp3a
Genomic Map