Assay Details
| Gene Symbol: kirrel3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099344.2 | NP_001092814.2 | 2 - 3 | 817 | 82 | |
| XM_021481418.1 | XP_021337093.1 | 2 - 3 | 853 | 82 | ||
| GenBank mRNA | CU638818.1 | - | 2 - 3 | 230 | 82 | |
| EF571006.1 | - | 2 - 3 | 817 | 82 | ||
More Information
Set Membership: |
|

kirrel3l
Genomic Map