Assay Details
| Gene Symbol: kirrel3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099344.2 | NP_001092814.2 | 6 - 7 | 1388 | 72 | |
| XM_021481418.1 | XP_021337093.1 | 6 - 7 | 1424 | 72 | ||
| GenBank mRNA | CU638818.1 | - | 6 - 7 | 801 | 72 | |
| EF571006.1 | - | 6 - 7 | 1388 | 72 | ||
More Information
Set Membership: |
|

kirrel3l
Genomic Map