Assay Details
| Gene Symbol: agpat9l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099450.1 | NP_001092920.1 | 10 - 11 | 1125 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

agpat9l| Gene Symbol: agpat9l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099450.1 | NP_001092920.1 | 10 - 11 | 1125 | 78 | |
Set Membership: |
|