Assay Details
| Gene Symbol: rplp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100436.2 | NP_001093906.1 | 2 - 3 | 180 | 72 | |
| GenBank mRNA | BC150391.1 | - | 2 - 3 | 184 | 72 | |
More Information
Set Membership: |
|

rplp2l
Genomic Map