Assay Details
| Gene Symbol: dad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100439.1 | NP_001093909.1 | 2 - 3 | 431 | 77 | |
| GenBank mRNA | BC150399.1 | - | 2 - 3 | 431 | 77 | |
More Information
Set Membership: |
|

dad1
Genomic Map