Assay Details
| Gene Symbol: plscr3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105113.1 | NP_001098583.1 | 3 - 4 | 194 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

plscr3a| Gene Symbol: plscr3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105113.1 | NP_001098583.1 | 3 - 4 | 194 | 127 | |
Set Membership: |
|