Assay Details
| Gene Symbol: senp3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105114.1 | NP_001098584.1 | 4 - 5 | 1156 | 90 | |
| XM_021476275.1 | XP_021331950.1 | 4 - 5 | 1181 | 90 | ||
More Information
Set Membership: |
|

senp3a
Genomic Map