Assay Details
| Gene Symbol: senp3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105114.1 | NP_001098584.1 | 5 - 6 | 1299 | 145 | |
| XM_021476275.1 | XP_021331950.1 | 5 - 6 | 1324 | 145 | ||
More Information
Set Membership: |
|

senp3a
Genomic Map