Assay Details
| Gene Symbol: hadhaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105276.1 | NP_001098746.1 | 18 - 19 | 2015 | 140 | |
| GenBank mRNA | BC115212.1 | - | 18 - 19 | 2015 | 140 | |
More Information
Set Membership: |
|

hadhaa
Genomic Map