Assay Details
| Gene Symbol: parvab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110471.1 | NP_001103941.1 | 6 - 7 | 724 | 68 | |
| GenBank mRNA | BC134924.1 | - | 6 - 7 | 724 | 68 | |
More Information
Set Membership: |
|

parvab
Genomic Map