Assay Details
| Gene Symbol: trpm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110479.1 | NP_001103949.1 | 4 - 5 | 576 | 97 | |
| XM_021470521.1 | XP_021326196.1 | 5 - 6 | 776 | 97 | ||
| XM_021470522.1 | XP_021326197.1 | 5 - 6 | 777 | 97 | ||
| GenBank mRNA | BC133947.1 | - | 4 - 5 | 576 | 97 | |
| KF305305.1 | - | 3 - 4 | 432 | 97 | ||
More Information
Set Membership: |
|

trpm1b
Genomic Map