Assay Details
| Gene Symbol: trpm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110479.1 | NP_001103949.1 | 5 - 6 | 702 | 71 | |
| XM_021470521.1 | XP_021326196.1 | 6 - 7 | 902 | 71 | ||
| XM_021470522.1 | XP_021326197.1 | 6 - 7 | 903 | 71 | ||
| GenBank mRNA | BC133947.1 | - | 5 - 6 | 702 | 71 | |
| KF305305.1 | - | 4 - 5 | 558 | 71 | ||
More Information
Set Membership: |
|

trpm1b
Genomic Map