Assay Details
| Gene Symbol: gdf6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130987.1 | NP_571062.1 | 1 - 2 | 459 | 99 | |
| GenBank mRNA | BC163352.1 | - | 1 - 2 | 540 | 99 | |
| BC163367.1 | - | 1 - 2 | 540 | 99 | ||
| X99769.1 | - | 459 | 99 | |||
More Information
Set Membership: |
|

gdf6b
Genomic Map