Assay Details
| Gene Symbol: fzd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130917.2 | NP_570992.2 | 1 | 1389 | 108 | |
| GenBank mRNA | AF117388.1 | - | 1 | 852 | 108 | |
| BC076546.1 | - | 1 | 1389 | 108 | ||
| BC165721.1 | - | 1 | 886 | 108 | ||
More Information
Set Membership: |
|

fzd10
Genomic Map