Assay Details
| Gene Symbol: krt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131156.2 | NP_571231.2 | 6 - 7 | 1268 | 77 | |
| XM_017353615.2 | XP_017209104.1 | 6 - 7 | 1306 | 77 | ||
| GenBank mRNA | AF197909.1 | - | 6 - 7 | 1260 | 77 | |
| BC063955.1 | - | 6 - 7 | 1268 | 77 | ||
| BC159195.1 | - | 4902 | 77 | |||
More Information
Set Membership: |
|

krt5
Genomic Map