Assay Details
| Gene Symbol: krt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131156.2 | NP_571231.2 | 7 - 8 | 1484 | 77 | |
| XM_017353615.2 | XP_017209104.1 | 7 - 8 | 1522 | 77 | ||
| GenBank mRNA | AF197909.1 | - | 7 - 8 | 1476 | 77 | |
| AJ131838.1 | - | 1 - 2 | 64 | 77 | ||
| BC063955.1 | - | 7 - 8 | 1484 | 77 | ||
| BC159195.1 | - | 5118 | 77 | |||
More Information
Set Membership: |
|

krt5
Genomic Map