Assay Details
| Gene Symbol: col2a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131292.1 | NP_571367.1 | 18 - 19 | 1178 | 119 | |
| XM_005166863.4 | XP_005166920.1 | 17 - 18 | 1110 | 119 | ||
| GenBank mRNA | BC163934.1 | - | 18 - 19 | 1170 | 119 | |
| BC167477.1 | - | 17 - 18 | 1074 | 119 | ||
| BC168504.1 | - | 18 - 19 | 1259 | 119 | ||
| DQ335127.1 | - | 18 - 19 | 1178 | 119 | ||
More Information
Set Membership: |
|

col2a1a
Genomic Map