Assay Details
| Gene Symbol: col2a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131292.1 | NP_571367.1 | 1 - 2 | 138 | 63 | |
| GenBank mRNA | BC163934.1 | - | 1 - 2 | 130 | 63 | |
| BC168504.1 | - | 1 - 2 | 219 | 63 | ||
| DQ335127.1 | - | 1 - 2 | 138 | 63 | ||
More Information
Set Membership: |
|

col2a1a
Genomic Map