Assay Details
| Gene Symbol: col2a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131292.1 | NP_571367.1 | 24 - 25 | 1640 | 57 | |
| XM_005166863.4 | XP_005166920.1 | 23 - 24 | 1572 | 57 | ||
| GenBank mRNA | BC163934.1 | - | 24 - 25 | 1632 | 57 | |
| BC167477.1 | - | 23 - 24 | 1536 | 57 | ||
| BC168504.1 | - | 24 - 25 | 1721 | 57 | ||
| DQ335127.1 | - | 24 - 25 | 1640 | 57 | ||
More Information
Set Membership: |
|

col2a1a
Genomic Map