Assay Details
| Gene Symbol: col2a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131292.1 | NP_571367.1 | 41 - 42 | 2792 | 101 | |
| XM_005166863.4 | XP_005166920.1 | 40 - 41 | 2724 | 101 | ||
| GenBank mRNA | BC163934.1 | - | 41 - 42 | 2784 | 101 | |
| BC167477.1 | - | 40 - 41 | 2688 | 101 | ||
| BC168504.1 | - | 41 - 42 | 2873 | 101 | ||
| DQ335127.1 | - | 41 - 42 | 2792 | 101 | ||
More Information
Set Membership: |
|

col2a1a
Genomic Map