Assay Details
| Gene Symbol: col2a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131292.1 | NP_571367.1 | 48 - 49 | 3491 | 113 | |
| XM_005166863.4 | XP_005166920.1 | 47 - 48 | 3423 | 113 | ||
| GenBank mRNA | BC163934.1 | - | 48 - 49 | 3483 | 113 | |
| BC167477.1 | - | 47 - 48 | 3387 | 113 | ||
| BC168504.1 | - | 48 - 49 | 3572 | 113 | ||
| DQ335127.1 | - | 48 - 49 | 3491 | 113 | ||
| U23822.1 | - | 5 - 6 | 366 | 113 | ||
More Information
Set Membership: |
|

col2a1a
Genomic Map