Assay Details
| Gene Symbol: col2a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131292.1 | NP_571367.1 | 53 - 54 | 4376 | 69 | |
| XM_005166863.4 | XP_005166920.1 | 52 - 53 | 4308 | 69 | ||
| GenBank mRNA | BC093433.1 | - | 3 - 4 | 635 | 69 | |
| BC163934.1 | - | 53 - 54 | 4368 | 69 | ||
| BC167477.1 | - | 52 - 53 | 4272 | 69 | ||
| BC168504.1 | - | 53 - 54 | 4457 | 69 | ||
| DQ335127.1 | - | 53 - 54 | 4376 | 69 | ||
| U23822.1 | - | 10 - 11 | 1251 | 69 | ||
More Information
Set Membership: |
|

col2a1a
Genomic Map