Assay Details
| Gene Symbol: elavl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131452.1 | NP_571527.1 | 5 - 6 | 706 | 79 | |
| XM_005171267.4 | XP_005171324.1 | 6 - 7 | 837 | 79 | ||
| XM_005171268.4 | XP_005171325.1 | 7 - 8 | 831 | 79 | ||
| XM_009298634.2 | XP_009296909.1 | 5 - 6 | 707 | 79 | ||
| XM_017357655.1 | XP_017213144.1 | 6 - 7 | 962 | 79 | ||
| GenBank mRNA | AF184244.1 | - | 5 - 6 | 706 | 79 | |
| BC044184.1 | - | 6 - 7 | 789 | 79 | ||
| U17600.1 | - | 3 - 4 | 413 | 79 | ||
More Information
Set Membership: |
|

elavl1a
Genomic Map