Assay Details
Gene Symbol: atp1a1a.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131689.1 | NP_571764.1 | 3 - 4 | 247 | 77 | |
GenBank mRNA | AY008376.1 | - | 3 - 4 | 266 | 77 | |
AY028629.1 | - | 3 - 4 | 247 | 77 | ||
BC054630.1 | - | 3 - 4 | 245 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
