Assay Details
| Gene Symbol: ncam1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131831.1 | NP_571906.1 | 19 - 20 | 3207 | 69 | |
| XM_005157405.4 | XP_005157462.1 | 23 - 24 | 3426 | 69 | ||
| XM_005157406.4 | XP_005157463.1 | 22 - 23 | 3402 | 69 | ||
| XM_005157407.4 | XP_005157464.1 | 21 - 22 | 3378 | 69 | ||
| XM_005157408.4 | XP_005157465.1 | 20 - 21 | 3375 | 69 | ||
| XM_005157409.4 | XP_005157466.1 | 21 - 22 | 3372 | 69 | ||
| XM_005157410.4 | XP_005157467.1 | 19 - 20 | 3350 | 69 | ||
| XM_005157411.4 | XP_005157468.1 | 18 - 19 | 3320 | 69 | ||
| XM_009291549.3 | XP_009289824.1 | 22 - 23 | 3423 | 69 | ||
| XM_009291550.3 | XP_009289825.1 | 22 - 23 | 3411 | 69 | ||
| XM_009291551.3 | XP_009289826.1 | 21 - 22 | 3408 | 69 | ||
| XM_009291552.3 | XP_009289827.1 | 22 - 23 | 3396 | 69 | ||
| XM_009291553.3 | XP_009289828.1 | 22 - 23 | 3393 | 69 | ||
| XM_009291554.3 | XP_009289829.1 | 21 - 22 | 3390 | 69 | ||
| XM_009291556.3 | XP_009289831.1 | 23 - 24 | 3012 | 69 | ||
| XM_009291559.3 | XP_009289834.1 | 19 - 20 | 3324 | 69 | ||
| XM_017359195.2 | XP_017214684.1 | 21 - 22 | 3387 | 69 | ||
| GenBank mRNA | AY029401.1 | - | 19 - 20 | 3207 | 69 | |
| BC162073.1 | - | 19 - 20 | 3107 | 69 | ||
| HM467819.1 | - | 19 - 20 | 2997 | 69 | ||
More Information
Set Membership: |
|

ncam1b
Genomic Map