Assay Details
| Gene Symbol: ncam1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131831.1 | NP_571906.1 | 9 - 10 | 1281 | 133 | |
| XM_005157405.4 | XP_005157462.1 | 10 - 11 | 1452 | 157 | ||
| XM_005157406.4 | XP_005157463.1 | 9 - 10 | 1428 | 133 | ||
| XM_005157407.4 | XP_005157464.1 | 10 - 11 | 1452 | 157 | ||
| XM_005157408.4 | XP_005157465.1 | 10 - 11 | 1452 | 157 | ||
| XM_005157410.4 | XP_005157467.1 | 9 - 10 | 1427 | 133 | ||
| XM_009291549.3 | XP_009289824.1 | 10 - 11 | 1452 | 157 | ||
| XM_009291550.3 | XP_009289825.1 | 10 - 11 | 1452 | 157 | ||
| XM_009291551.3 | XP_009289826.1 | 10 - 11 | 1452 | 157 | ||
| XM_009291553.3 | XP_009289828.1 | 10 - 11 | 1452 | 157 | ||
| XM_009291554.3 | XP_009289829.1 | 10 - 11 | 1452 | 157 | ||
| XM_009291556.3 | XP_009289831.1 | 10 - 11 | 1452 | 157 | ||
| XM_009291557.3 | XP_009289832.1 | 10 - 11 | 1452 | 157 | ||
| XM_009291558.3 | XP_009289833.1 | 10 - 11 | 1452 | 157 | ||
| XM_017359195.2 | XP_017214684.1 | 9 - 10 | 1428 | 133 | ||
| GenBank mRNA | AY029401.1 | - | 9 - 10 | 1281 | 133 | |
| BC162073.1 | - | 9 - 10 | 1181 | 133 | ||
| CU458747.1 | - | 9 - 10 | 1071 | 133 | ||
| CU458882.1 | - | 10 - 11 | 1095 | 157 | ||
| HM467819.1 | - | 9 - 10 | 1071 | 133 | ||
More Information
Set Membership: |
|

ncam1b
Genomic Map